Would you like to log in again?
Monoject™ Medication Transfer Needle, Covidien
Supplier: Covidien
Monoject™
8881204005
15426-168CS 611.01 USD InStock
15426-168
Monoject™ Medication Transfer Needle, Covidien
Syringe Needles
Double-ended, 20-gauge, stainless steel needle with special anti-coring bevels
Features a smooth, rounded aluminum hub and a short butt end for evacuation. Sterile.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty