Would you like to log in again?
Dispensing Bottle, HDPE, Round, Menda
Supplier: Menda
35608 35601
25445-029EA 15 USD InStock
25445-029 15431-356
Dispensing Bottle, HDPE, Round, Menda
Bottles Dropping and Dispensing Bottles
Dispensing bottle is made from high denisty polyethylene/amber glass.
  • Durable
  • Alcohol imprint for demarcation
  • Lasting touch pump
Ideal for alcohol dispensing. Amber glass construction protects light and plastic sensitive fluids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty