Would you like to log in again?
Dispensing Bottles, HDPE, Square, Menda
Supplier: Menda
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
35312 35505 35508 35309 35703 35702 35305 35704
15431-722EA 28.82 USD InStock
15431-722 15431-744 15431-706 15431-726 15431-354 15431-742 15431-358 15431-740
Dispensing Bottles, HDPE, Square, Menda
Bottles Dropping and Dispensing Bottles
Dispensing bottle suitable for many solvents.
  • Wide opening, easy to fill
  • Provides stability and saves storage space
  • Translucent for easy viewing of contents
Dispenses approximately 0.2 cc of liquid with each pump.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty