Would you like to log in again?
One-Touch Pump, Menda
Supplier: Menda
35300
15431-758EA 24.77 USD InStock
15431-758
One-Touch Pump, Menda
Pumps Vacuum Pumps
One-Touch pumps are made of precision machined valves for smooth consistent operation that also eliminates spraying and splashing.
  • Cam-lock hinge keeps lid open and closed that reduces odors
  • Fits all Standard Menda HDPE, glass, and steel bottles
  • Dispenses approximately 0.2 cc of liquid with each pump
  • Silicone and latex free that are perfect for ultra-clean environments
  • Not recommended for use with acrylic liquid
  • Made of stainless steel
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty