Would you like to log in again?
Small Nut Starter, Desco Industries
Supplier: Desco Industries
35120
15431-818EA 3.71 USD InStock
15431-818
Small Nut Starter, Desco Industries
Nutdrivers
The small nut starter helps to hold and install small mounting hardware.
  • 4" length
  • Polyethylene (LDPE) construction
Holds nut sizes regular no. 2 through small no. 6. US nut size dimensions ranging from 3/16 to 5/16". Metric size dimensions ranging from 4 to 10 mm. Made in the United States of America for superior quality.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty