Would you like to log in again?
AQUALOCKIT IN 2# SHAKER BOTTL
Supplier: FYTERTECH NONWOVENS LLC
2# AQUALOCKIT* Shaker Bottle^ AQUALOCKIT* is ideal for easy and effective clean-up of contaminated water, sludge, neutralized acids or bases or low-level radioactive water.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
AQUALOCKIT IN 2# SHAKER BOTTL 15551-588 670001 Case of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty