Would you like to log in again?
ROLL 16X150' HAZMAT DW CS2
Supplier: FYTERTECH NONWOVENS LLC
Spilfyter* Hazmat Roll^ 40.6cmx45.7m double weight perforated roll. Absorbs up to 16 times its weight. Quick "No-Decision Response" for unknown spills. Absorbs most aggressive and non-aggressive liquids. Yellow-colored sorbents alert of potential danger. Made with smaller micron diameter fibers.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ROLL 16X150' HAZMAT DW CS2 15551-644 S2-91 Case of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty