Would you like to log in again?
ROLL 32X300' HAZMAT SW
Supplier: FYTERTECH NONWOVENS LLC
Hazmat Sorbent Roll^ Standard weight. Dimensions: 81.2cm x 91.4m (32"x300'). Excellent liquid retention, capillary action disperses liquid as its absorbed. Absorbs aggressive and nonaggressive liquids. Yellow color provides visual alert of potential danger. Horizontally perforated every 61cm (24")
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ROLL 32X300' HAZMAT SW 15551-646 S2-95 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty