Would you like to log in again?
ROLL 18X150' CELLULOSE
Supplier: FYTERTECH NONWOVENS LLC
18in.x150ft. Cellulose Roll^ Spilfyter* Cellulose-Based Rolls are reinforced with a top and bottom liner for increased durability. Easily absorbs oil, coolants, solvents and water.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ROLL 18X150' CELLULOSE 15551-668 151048 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty