Would you like to log in again?
SPILFYTER® Sorbents for Hydrocarbons, NPS
Supplier: NPS
SPILFYTER®
M-37 M-75 M-55
15551-746CS 220.8 USD InStock
15551-746 15551-692 15551-880
SPILFYTER® Sorbents for Hydrocarbons, NPS
Spill Control Materials
For quick absorption of hydrocarbon spills
Sock is designed for quick, easy oil pick-up. The superior absorbency and wicking action make this sock especially suitable for recovering oil spills on land.
Melt-blown pad is ideal for collection of lightweight hydrocarbons such as J P-4 fuel and kerosene.
Boom is designed to handle easily on water. Absorbs oils quickly. The sealed polypropylene case makes the boom environmentally safe.
SPILFYTER® Sorbents are made of 100% polypropylene fabric that repels water while attracting and absorbing oils and hydrocarbon-based fuels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty