Would you like to log in again?
SPILFYTER® Hazmat Pad, NPS
Supplier: NPS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SPILFYTER®
S2-70 S2-71 S2-75
56609-392CS 374.08 USD InStock
56609-392 15551-702 56609-396
SPILFYTER® Hazmat Pad, NPS
Spill Control Materials
Single-weight perforated pad absorbs up to 16 times its own weight in virtually all liquids.
  • Absorbs up to 16X its own weight in virtually all liquids, aggressive or nonaggressive
  • Perforated, allows quick response when nature of spill is unknown
  • Pad is constructed of smaller diameter fibers providing greater liquid retention and strength
  • Yellow color alerts others of potential danger
Single-weight perforated pad absorbs up to 16 times its own weight in virtually all liquids, either aggressive or nonaggressive, and allows a quick response when the nature of the spill is unknown. Pad is constructed of smaller diameter fibers that provide greater liquid retention and strength. Yellow color alerts others of potential danger.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty