Would you like to log in again?
SPILL KIT 170 GALLON HAZMAT CA
Supplier: FYTERTECH NONWOVENS LLC
Hazmat 170-gallon Spill Cart^ Absorbs 135 gallons. Contents: (40) 3in.x4ft. socks, (26) 18in.x18in. socks, (200) 16in.x18in. pads, (15) 18in.x18in. pillows, (1) 16in.x150ft. roll, (70) wet wipes, (2pr.) gloves, (2pr.) goggles, (15) disposal bags, (1) emergency guidebook, (1) wheeled dispenser cart.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPILL KIT 170 GALLON HAZMAT CA 15551-806 250170 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty