Would you like to log in again?
SPILL KIT GRAB + GO OIL ONLY
Supplier: FYTERTECH NONWOVENS LLC
Oil-Only Grab & Go* Portable Spill Kit^ Absorbs 4 gallons. Contains all types of non-aggressive liquid drips, leaks and spills before migration occurs. Contents: (4) 3in.x4ft. socks, (3) 16in.x18in. premium pads, nitrile gloves, goggles, 5-gallon bucket w/snap on lid.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPILL KIT GRAB + GO OIL ONLY 15551-814 305304 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty