Would you like to log in again?
SPILL KIT 55 GALLON OIL ONLY
Supplier: FYTERTECH NONWOVENS LLC
Oil-Only Poly Drum Spill Kit^ Absorbs up to 40 gallons. Contents: (20) 3in.x4ft. socks, (200) 16in.x18in. pads, (1) disposal bag & twist tie, nitrle gloves, (1pr.) splash goggles, (1) 55-gallon poly drum.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPILL KIT 55 GALLON OIL ONLY 15551-822 322055 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty