Would you like to log in again?
SPILL KIT 95 GALLON OIL ONLY
Supplier: FYTERTECH NONWOVENS LLC
Oil-Only 95 Gallon Overpack Spill Kit^ Absorbs up to 82 Gallons. Contents: (40) 3in.x4ft. socks, (200) 16x18in. pads, (10) 18x18in. pillows, (4) 3in.x10ft. socks, (1) 8in.x20ft. boom, (5) disposal bags, (2pr.) nitrile gloves, (2pr.) splash goggles, (1) 95-gallon overpack drum w/screw top lid.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPILL KIT 95 GALLON OIL ONLY 15551-824 350095 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty