Would you like to log in again?
SPILFYTER® Mercsorb® Mercury Amalgamation Powder, NPS
Supplier: NPS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
520500
15551-838EA 484.96 USD InStock
15551-838
SPILFYTER® Mercsorb® Mercury Amalgamation Powder, NPS
Spill Control Materials
This Mercury Amalgamation Powder is used to convert elemental mercury into an amalgam, which stops dangerous mercury vapors from being emitted.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty