Would you like to log in again?
PILLOWS 10X10 CELLULOSE CS12
Supplier: FYTERTECH NONWOVENS LLC
10x10in. Cellulose Pillow^ Cellulose-based pillows are primarily for non-aggressive spills. Use to absorb leaks in small or confined spaces.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PILLOWS 10X10 CELLULOSE CS12 15552-150 150032 Case of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty