Would you like to log in again?
SPILFYTER® Chemical Classifier Strips, NPS
Supplier: NPS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SPILFYTER®
570010 570050 570001 577777
15552-166CS 300.1 USD InStock
15552-166 56609-230 56616-372 56609-254
SPILFYTER® Chemical Classifier Strips, NPS
Spill Control Materials
Distinguish chemical risks in unknown spilled liquids and classify for proper treatment in a matter of seconds.
  • Strips simultaneously test for acid/base, oxidizer, fluoride, petroleum product/organic solvent, and iodine/bromine/chlorine risks
  • Laminated chemical classifier chart determines possible presence of chemical types according to color changes in strip
Each chemical classifier station includes 20 chemical classifier strips, one chemical classifier chart, two bobbins of polypropylene thread, and one weather-resistant plastic carry case. Each chemical classifier kit includes six chemical classifier strips, one chemical classifier chart, and one bobbin of polypropylene thread. Additional strips and charts are available separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty