Would you like to log in again?
Spilfyter® Waste Water Classifier, J.V. Mfg.
Supplier: NPS
SPILFYTER®
580050 581020 580001 580010
15552-170CS 284.73 USD InStock
15552-170 15552-172 15552-174 56609-232
Spilfyter® Waste Water Classifier, J.V. Mfg.
Environmental Test Kits
Distinguish risks in unknown wastewater batches and classify for proper treatment in a mater of seconds
Each wastewater classifier station (15552-174) includes 20 wastewater classifier strips, one wastewater classifier chart, two bobbins of polypropylene thread, and one weather-resistant plastic carry case. Each wastewater classifier kit (56609-232) includes six wastewater classifier strips, one wastewater classifier chart, and one bobbin of polypropylene thread. Additional strips and charts are available separately.
Strips simultaneously test for acid/base, petroleum/organic solvent, hydrogen sulfide, nitrite, nitrate, and fluoride risks. Laminated wastewater classifier chart determines possible presence of wastewater chemical types according to color changes in strip.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty