Would you like to log in again?
SPILFYTER® All-in-One Sorbents, NPS
Supplier: NPS
SPILFYTER®
DB-914 Z-914
15552-234EA 259.15 USD InStock
15552-234 15552-236
SPILFYTER® All-in-One Sorbents, NPS
Spill Control Materials
Unique fine fiber sorbents absorb up to 16 times their own weight and can be used indoors or outdoors as a pad for small localized spills, a sock for wrapping around machinery and equipment, a wipe for cleaning tools or small spills, or a roll for large coverage areas
Sorbent 15552-236 is for oil only and contains small micron diameter fibers which provide greater liquid retention and strength. The oil-only sorbent can float while absorbing oil and other hydrocarbons and repelling water.
Sorbents are reinforced on the top layer for added durability, perforated horizontally every 30.5cm (12"), and vertically zipped for easy folding. Dimensions: 40.6Wcmx45.7Lm (16"x150').
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty