Would you like to log in again?
Super Sani-Cloth® Germicidal Disposable Wipes, PDI®
Supplier: PDI
Super Sani-Cloth®
PDIQ86984 PDIU87295 PDIQ55172 PDIH04082
15648-962CS 280.12 USD InStock
15648-962 15648-993 10799-096 10799-098
Super Sani-Cloth® Germicidal Disposable Wipes, PDI®
Wipes Disposable Wipes and Towels Disinfectant Wipes
These disposable wipes are saturated with a quaternary/high alcohol formula for the disinfection of hard, non-porous surfaces.
They are tested effective against 26 microorganisms, and can kill TB and RSV in 1 minute and H1N1, Rotavirus, MRSA, VRE, and ESBL Producing E. coli in 2 min. The distinctive hexagonal-embossed material contains 40% wood-based fibers for a more cloth-like feel.
Ordering information: Each extra-large canister contains 65 wipes; case contains six canisters. The individually wrapped wipes are packaged 50 per box; case contains 3 boxes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty