Would you like to log in again?
SharpSafety™ In-Room™ System Wall Enclosure, Covidien
Supplier: Covidien
SharpSafety™ In-Room™ In-Room™
85161H
15704-170CS 167.2 USD InStock
15704-170
SharpSafety™ In-Room™ System Wall Enclosure, Covidien
Bins Waste Bins
Locked ABS-plastic enclosure secures a 1.9 L (2 qt.) or 4.7 L (5 qt.) sharps-disposal container for a secondary barrier of protection.
  • Locked Sharps-Container Mounting
  • Secure Point-of-Use Disposal
  • Long-Lasting ABS Plastic Provides a Secondary Barrier of Protection
  • Viewing Windows on Front and Sides
  • Beige, Unobtrusive Style Fits Any Room Decor
Viewing windows on front and sides make it easy to monitor the container fill level.
This wall enclosure has a variety of convenient features. All cabinets are universally keyed, and the viewing windows on the front and sides make monitoring the fill level easy.
Certifications: Meets or exceeds JCAHO, OSHA, and NIOSH/CDC standards and requirements.
Ordering information: For sharps containers, see the In-Room Sharps Disposal Systems series from Covidien.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty