Would you like to log in again?
Sharps Disposal Containers with Horizontal-Drop Opening Lids, Covidien
Supplier: Covidien
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
89671 89651
15704-200CS 363.94 USD InStock
15704-200 15704-236
Sharps Disposal Containers with Horizontal-Drop Opening Lids, Covidien
Bins Sharps Containers
Clear lid with large horizontal opening accommodates larger syringes, allowing one-hand, one-step, no-touch sharps depositing
Anti-spill inner flap on the lid impedes hand entry. Container base has hand grips for easy transport. Containers lock for final disposal. Plastics and pigments are safe to autoclave or incinerate. Containers can be placed on countertops or mounted on a wall.
Certifications: Containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty