Would you like to log in again?
Jar, Polypropylene, White, Wide Mouth, Qorpak®
Supplier: Qorpak
PLA-03349 PLC-07186 PLC-07184 PLC-07189 PLC-07188 PLC-07185 PLC-07187
11311-350CS 98.02 USD InStock
11311-350 11311-352 15712-134 11311-348 11311-358 11311-354 11311-356
Jar, Polypropylene, White, Wide Mouth, Qorpak®
Bottles Plastic Bottles
White polypropylene jars are ideal for solids, powders and cosmetics.
Jars are ideal for storing and collecting specimens and lab supplies. Wide neck and straight sides facilitate easy recovery of contents. Polypropylene offers greater tolerance to high temperatures than HDPE and better contact clarity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty