Would you like to log in again?
Nalgene® Fluorinated High-Density Polyethylene, Wide-Mouth Bottles, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2197-0016 2197-0004 2197-0008 2197-0032
15712-944CS 474.24 USD InStock
15712-944 15712-946 15712-940 15712-942
Nalgene® Fluorinated High-Density Polyethylene, Wide-Mouth Bottles, Thermo Scientific
Bottles Plastic Bottles
Nalgene® Fluorinated High-Density Polyethylene Wide-Mouth Bottles feature a fluorinated surface that provides improved barrier properties and reduces solvent absorption and permeation.
  • Bottles are a safe, durable, cost-efficient line and the fluorination enhances long-term container performance
  • Prevents material loss due to permeation and yields lower extractables
  • Wide-mouth design allows for easy filling and dispensing of solid and liquid materials
  • Bottles are split- and puncture-resistant, as well as recyclable
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty