Would you like to log in again?
Urinometer, Profex Medical Products
Supplier: Profex Medical Products
760
15744-325CS 1266.65 USD InStock
15744-325
Urinometer, Profex Medical Products
Clinical Diagnostic Supplies Clinical Instruments
Glass urinometer has a reinforced, clear, heavy fluted jar with extra-wide base and pouring tip
  • Specific gravity range: 1.000 to 1.060
Shot-weighted hydrometer float.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty