Would you like to log in again?
Quick Disconnect Coupling, APC Series ¼" Flow, Colder Products Company
Supplier: CPC (Colder Products Company)
APCD17004SH APC22004 APC17006SH APC17004 APCD17006SH APC22006 APC23004 APC10006 APCD10004 APC17004SH APCD22004 APC23006 APCD23006 APCD22006 APC24004 APC17006 APC10004
16001-368PK 36.73 USD InStock
16001-368 16001-358 16001-386 16001-354 16001-376 16001-366 16001-378 16001-356 16001-382 16001-360 16001-372 16001-362 16001-384 16001-374 16001-352 16001-380 16001-370
Quick Disconnect Coupling, APC Series ¼" Flow, Colder Products Company
Fittings and Connectors
The 1/4" Flow APC series coupling features a plastic thumb latch, fewer moving parts and a smooth contoured design to deliver excelent flow in a compact size
Materials of construction include Acetal, 316 and 302 stainless steel, Buna-N o-ring seal.
The shrouded thumb latch protects against accidental disconnect.
Ordering information: Accessories not included
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty