Would you like to log in again?
CQH ChemQuik Series Moderate Flow, Quick Disconnect Coupling, Colder Products Company
Supplier: CPC (Colder Products Company)
CQHD06200206 CQHD06200104 CQHD06110104 CQHD06110106 CQHD06100108 CQHD06200108 CQHD06100106 CQHD06110206 CQHD06100206 CQHD06100104
16001-402PK 115.03 USD InStock
16001-402 16001-398 16001-388 16001-400 16001-394 16001-396 16001-406 16001-390 16001-408 16001-392
CQH ChemQuik Series Moderate Flow, Quick Disconnect Coupling, Colder Products Company
Fittings and Connectors
These 1/2" pressure balanced, non-spill Quick Disconnect Couplings are intended for high purity and aggressive chemical applications
Materials of construction include natural polypropylene or PVDF, wetted PEEK spring, FKM and FFKM o-ring seal options.
A non-metal flow path allows for a braod range of chemical compatibilities.
Ordering information: Accessories not included
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty