Would you like to log in again?
CQG ChemQuik Series 3/8" Flow, Quick Disconnect Coupling, Colder Products Company
Supplier: CPC (Colder Products Company)
CQGD06200112 CQGD06100208 CQGD06100112 CQGD06100108 CQGD06200106 CQGD06200208 CQGD06200108 CQGD06100106
16001-434PK 234.3 USD InStock
16001-434 16001-446 16001-436 16001-442 16001-432 16001-444 16001-440 16001-438
CQG ChemQuik Series 3/8" Flow, Quick Disconnect Coupling, Colder Products Company
Fittings and Connectors
These 3/8" pressure balanced, non-spill Quick Disconnect Couplings are intended for high purity and aggressive chemical applications
Materials of construction include natural polypropylene, non-wetted Hastelloy C spring, FKM and FFKM o-ring seal options.
A non-metal flow path allows for a braod range of chemical compatibilities.
Ordering information: Accessories not included
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty