Would you like to log in again?
HFC39 Series Plastic Couplings, Colder Products Company
Supplier: CPC (Colder Products Company)
HFC22439M HFCD22839M HFC22839M HFCD17639M HFCD22439M HFCD22639M HFCD17439M HFCD17839M
16001-456PK 440.97 USD InStock
16001-456 16001-458 16001-448 16001-464 16001-454 16001-460 16001-450 16001-462
HFC39 Series Plastic Couplings, Colder Products Company
Fittings and Connectors
These high-flow couplings feature automatic shut-off valves which stop flow and eliminate the need for pinch clamps, stainless steel connectors, or fittings
They allow rapid disconnect and reconnect without the need for tools. Valved couplings stop fluid flow at the point of connection, reducing spills and related safety issues. Couplings feature an audible click when engaged, ensuring the user of a secure connection. Constructed of medical-grade polysulfone (amber tint) with clear silicone seals. Couplings are sterilizable, reusable, and compatible with virtually all existing processes.
Certifications: All products meet Class VI criteria and feature BSE/TSE-free materials (ADCF).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty