These 1/4" flow chrome plated brass quick disconnect couplings are built to withstand the most demanding applications and retain their attractive, refletive appearance
Materials of construction includes chrom plated brass, 316 stainless steel, acetal, Buna N oring seals.
For design flexibility these couplings are available in an NSF listed version and can mate with PLC couplings as well.
Ordering information: Accessories not included