Would you like to log in again?
Chrome Plated Brass LC Series 1/4" Flow, Quick Disconnect Coupling, Colder Products Company
Supplier: CPC (Colder Products Company)
LCD17006 LCD23006 LC21006 LC10006 LC13006 LC42006 LCD40004 LC22005 LCD16006 LC24004 LCD16005 LC10004 LC40006 LCD42005 LC22006 LC22004 LC17004 LC20006 LC42004 LC15004 LCD13006 LCD22006 LC17005 LC20004 LCD40006 LC17006 LC21004 LCD10004 LC12006 LCD10006 LCD13004 LCD17004 LC12004 LCD17005 LC16004 LCD24006 LCD22004 LC40004 LCD24004 LCD15004 LC23004 LC16005 LCD12004 LC26004 LCD22005 LC23006 LCD42004 LCD12006 LCD23004 LC16006 LCD20004 LCD21004 LC42005 LC13004 LCD20006 LCD42006
16001-578PK 283.25 USD InStock
16001-578 16001-534 16001-536 16001-496 16001-530 16001-498 16001-576 16001-532 16001-538 16001-492 16001-570 16001-494 16001-572 16001-490 16001-566 16001-522 16001-568 16001-524 16001-562 16001-486 16001-520 16001-564 16001-488 16001-526 16001-528 16001-482 16001-560 16001-484 16001-480 16001-478 16001-556 16001-514 16001-474 16001-552 16001-476 16001-510 16001-554 16001-516 16001-518 16001-470 16001-472 16001-550 16001-544 16001-500 16001-468 16001-502 16001-546 16001-584 16001-542 16001-466 16001-508 16001-504 16001-548 16001-506 16001-580 16001-582
Chrome Plated Brass LC Series 1/4" Flow, Quick Disconnect Coupling, Colder Products Company
Fittings and Connectors
These 1/4" flow chrome plated brass quick disconnect couplings are built to withstand the most demanding applications and retain their attractive, refletive appearance
Materials of construction includes chrom plated brass, 316 stainless steel, acetal, Buna N oring seals.
For design flexibility these couplings are available in an NSF listed version and can mate with PLC couplings as well.
Ordering information: Accessories not included
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty