Would you like to log in again?
MPX Series 1/2" Flow, Quick Disconnect Coupling for Bio and Pharma, Colder Products Company
Supplier: CPC (Colder Products Company)
MPX17839 MPXK17839 MPX22803M MPX22839M MPXK17803 MPX17803
16001-632PK 4281.28 USD InStock
16001-632 16001-634 16001-630 16001-636 16001-626 16001-628
MPX Series 1/2" Flow, Quick Disconnect Coupling for Bio and Pharma, Colder Products Company
Fittings and Connectors
These 1/2" flow quick disconnect couplings are manufactured specifically for the bio and pharma markets
Materials of construction include animal free (BSE/TSE) USP class 6 polysulfone and polycarbonate, Buna - n and platinum cured USP class 6 silicone oring seals.
USP Class 6 and animal free (BSE/TSE) materials are appropriate for the Bio, Pharma and Life Sciences with single use dispoability in mind.
Certifications: USP Class VI, BSE/TSE free materials (ADCF)
Ordering information: Accessories not included
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty