1/4" flow quick disconnect couplings available in either polypropelene or acetal offer good chemical resistance for most applications
Materials of construction include polypropylene or acetal, 316 stainless steel and Buna N or EPDM oring seals.
Availabe with NSF listed materials and can mate with LC seried connectors for design flexibility.
Ordering information: Accessories not included