Would you like to log in again?
PLC Series 1/4" Flow, Quick Disconnect Coupling, Colder Products Company
Supplier: CPC (Colder Products Company)
PLCD29006 PLC1000412 PLCD1300612 PLC14004 PLCD41004 PLC42005 PLC2400412 PLCD40006 PLCD23004 PLCD1700412 PLCD1700612 PLCD2000612 PLCD11004 PLCD2300412 PLCD14004 PLC23006 PLC16005 PLC1600612 PLCD1000612 PLC29006 PLC2200612 PLCD2300612 PLC2300612 PLC1700612 PLCD2400412 PLCD17005 PLC2300412 PLCD2200612 PLCD2200412 PLC29004 PLC21006 PLCD16005 PLC23004 PLCD1000412 PLCD1200612 PLC22005 PLCD23006 PLCD22005 PLCD14006 PLCD4200412 PLCD1600412 PLC1600412 PLC2200412 PLCD21006
16001-698PK 158.57 USD InStock
16001-698 16001-676 16001-710 16001-798 16001-754 16001-776 16001-656 16001-734 16001-778 16001-694 16001-794 16001-772 16001-652 16001-752 16001-658 16001-758 16001-736 16001-716 16001-738 16001-690 16001-692 16001-670 16001-770 16001-764 16001-720 16001-742 16001-688 16001-722 16001-788 16001-744 16001-766 16001-800 16001-782 16001-640 16001-662 16001-784 16001-686 16001-728 16001-702 16001-704 16001-804 16001-680 16001-780 16001-682
PLC Series 1/4" Flow, Quick Disconnect Coupling, Colder Products Company
Fittings and Connectors
1/4" flow quick disconnect couplings available in either polypropelene or acetal offer good chemical resistance for most applications
Materials of construction include polypropylene or acetal, 316 stainless steel and Buna N or EPDM oring seals.
Availabe with NSF listed materials and can mate with LC seried connectors for design flexibility.
Ordering information: Accessories not included
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty