Would you like to log in again?
PLC Series Plastic Couplings, Colder Products Company
Supplier: CPC (Colder Products Company)
PLCD20006 PLC10006 PLC20004 PLC12004 PLC22004 PLCD13004 PLCD17006 PLC24004 PLCD20004 PLC40004 PLCD22004 PLC17006 PLCD12006 PLCD42006 PLC16006 PLC42004 PLC13006 PLCD12004 PLC20006 PLC42006 PLCD16006 PLCD22006 PLCD40004 PLC22006 PLC16004 PLCD10004 PLCD17004 PLCD16004 PLCD24006 PLCD24004 PLCD10006 PLC24006 PLC17004 PLC13004
16001-654PK 315.57 USD InStock
16001-654 16001-678 16001-756 16001-712 16001-672 16001-650 16001-750 16001-696 16001-674 16001-730 16001-774 16001-796 16001-718 16001-714 16001-790 16001-786 16001-644 16001-700 16001-668 16001-760 16001-684 16001-762 16001-740 16001-642 16001-664 16001-706 16001-806 16001-708 16001-768 16001-724 16001-746 16001-648 16001-726 16001-660
PLC Series Plastic Couplings, Colder Products Company
Fittings and Connectors
These quick disconnect couplings are injection molded from acetal thermoplastic, making them highly durable and resistant to most chemical solutions
Both the external and valve springs are constructed of stainless steel. Couplings feature integral terminations which results in fewer leak points, shorter assemblies, and faster installations. An audible click assures the user of a secure and reliable connection. Couplings have a thumb latch for rapid disconnect and reconnect with one hand, without the need for tools. Valved couplings stop fluid flow at the point of connection, reducing spills and related safety issues.
Ordering information: Polypropylene couplings for greater chemical resistance are available; contact your Avantor sales representative.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty