1/16" flow miniature quick disconnect couplings provide a reliable and secure alternative to luer type fittings
Materials of construction include polypropylene, acetal and chrome plated brass, 316 stainless steel springs and eithe EPDM or Buna N oring seals.
Unique design allows tubingto rotate preventing kinks and accidental disconnections.
Ordering information: Accessories not included