Would you like to log in again?
Tissue Section Adhesive, Epredia™, Richard-Allan Scientific
Supplier: Richard-Allan Scientific
Epredia™
86014
16002-126EA 107.6 USD InStock
16002-126
Tissue Section Adhesive, Epredia™, Richard-Allan Scientific
Stains Microscopy and staining reagents
Increase the affinity of the tissue for the slide with Epredia™ Tissue Section Adhesive.
  • Use in the water bath to increase the affinity of the tissue for the slide
  • Creates no background residue
  • Improves adhesion for techniques utilizing heat
Pre-mixed and ready-to-use, this adhesive will not interfere with staining reactions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty