Would you like to log in again?
Epredia™ Microscope Slides and Cover Glasses
Supplier: Epredia
6776214 202460 9991001 9991011 4446 102450 102250 4448 4443 152250 152440 152455 4450 202450 152222 12450S 9991003 12460S 4442 9991004 12450S 9991002 152460 12460S 4451 102260 4444 9991014 102440 102222 9991095 152450 9991009 9991012 152450 152222 102424 12440S 4441 9991013 6776214CS 4447 202450 102240 152440 202460 4445 102460 152250 152455 12440S 102460 9991090 102455 4440 152460 102430
10118-578CS 47.46 USD InStock
10118-578 10144-820 10118-576 16002-262 16002-266 10118-582 16002-264 10118-580 16002-268 10118-584 CA16002-316 CA16002-314 10118-568 10118-566 10118-962 10118-570 16002-234 16002-314 10118-574 16002-238 10118-572 16002-236 16002-316 10118-958 10034-100 10118-952 10118-950 16002-240 10118-956 10118-954 16002-244 16002-242 CA16002-252 16002-248 CA16002-532 10118-960 CA16002-258 16002-246 CA16002-530 CA16002-256 CA16002-534 10034-098 10118-948 10118-942 10118-946 16002-252 10118-944 16002-250 CA16002-264 16002-530 16002-256 CA16002-262 16002-534 CA16002-268 16002-532 CA16002-266 16002-258
Epredia™ Microscope Slides and Cover Glasses
Slides Microscope Slides
Epredia™ offers a wide assortment of high-quality microscope slides and cover glass sizes that are available to meet the needs of modern laboratories.
  • Epredia™ cover glass and coverslips are uniquely manufactured to eliminate sticking and reduce waste
  • Made from the highest quality glass
  • Free from blisters and flaws, and consistent in cut, thickness and dimensional accuracy
  • Signature Series™ cover glass are an excellent choice for use with automatic coverslippers
Superfrost™ Plus and Colorfrost Plus™ microscope slides are positively charged for improved cellular adhesion. They electrostatically attract tissue sections and cytology preparations and bind them to the slide. This eliminates the need for special adhesives or protein coatings, thus avoiding background staining. These slides are available in 10 different colors.
Permafrost™ microscope slides are available in a wide variety of colors that are resistant to commonly used laboratory solvents, making them an excellent choice for all laboratory applications. This allows users to easily sort workload by color tab. These slides are available in 11 different colors.
Epredia Multi-Spot™ Slides feature a coating that enables the slides to undergo multiple washings for reuse. They feature either 6mm or 9 mm diameter wells, with a total of 12 wells per slide. Wells are surrounded by a chemical- and water-resistant PTFE (polytetrafluoroethylene) coating.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty