Would you like to log in again?
Heating Mantles, Upper Portion for Unjacketed Cylindrical Vessels, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-10013-50 CG-10013-10 CG-10013-30 CG-10013-20 CG-10013-21 CG-10013-15
16002-574EA 3270.71 USD InStock
16002-574 16002-584 16002-578 16002-576 16002-582 16002-580
Heating Mantles, Upper Portion for Unjacketed Cylindrical Vessels, Chemglass
Heaters Heating Mantles
Glass fabric heating mantle with a lengthwise slit, held together with a hook and lace closure.
  • For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels
  • 115 volts
Delivery information: Supplied complete with a separable 4', 2-wire cord and locking connector.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty