Would you like to log in again?
Flow Control Adapters, 90°, with PTFE Stopcock, Chemglass
Supplier: Chemglass
CG-1030-19 CG-1030-29 CG-1030-45
80065-910EA 148.39 USD InStock
80065-910 80065-912 16002-604
Flow Control Adapters, 90°, with PTFE Stopcock, Chemglass
Adapters Adapters, Joints with Tube Connectors
Used to regulate the flow of gases or liquids into reaction systems.
  • Equipped with a PTFE stopcock
  • For replacement stopcocks, see CG-424-77
  • 90° angled
Hose connection has an O.D. of 8 mm at the middle serration with the exception of CG-1030-45, which has an O.D. of 14 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty