Would you like to log in again?
Thermocouple Adapters, Chemglass
Supplier: Chemglass
CG-1042-E-07 CG-1042-E-02 CG-1042-E-10 CG-1042-E-01 CG-1042-E-06
16002-630EA 112.38 USD InStock
16002-630 16002-634 16002-626 16002-624 16002-628
Thermocouple Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters feature a Chem-Thread at the top to make a vacuum tight seal.
  • Accepts 1/8" or 1/4" O.D. thermocouple probes
Lower portion of the adapter is reduced down to hold the thermocouple securely in place and away from the agitators.
Delivery information: Supplied complete with a Celcon cap and Viton O-Ring. Perfluoro O-Rings are also available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty