Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buchner Funnels, Table Top, PE, Porous, 18", Chemglass

Supplier: Chemglass
CG-1413-07 CG-1413-06 CG-1413-05 CG-1413-08
16002-720EA 1969.33 USD InStock
16002-720 16002-714 16002-718 16002-716
Buchner Funnels, Table Top, PE, Porous, 18", Chemglass
Funnels Filtration Funnels Büchner Funnels
Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate.

  • Removable hose barb connector accepts 12.7 mm (¹/₂") Int.Ø tube and threads into 3/4" NPT bung on the funnel
  • Fixed or removable filter plates are supported by a multiple ring grid
  • HDPE, 6.4 mm (¹/₄") thick, with a non-porous ring at the periphery of the plate which seals filter paper when used with porous filter plates
  • Use temperature range maximum 52 °C (125 °F), minimum –20 °C (–4 °F)

Available in two porosities (medium and coarse); medium is 45 to 90 µm while the coarse is 90 to 130 microns.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty