Would you like to log in again?
Accessories for Reaction Vessels, Chemglass
Supplier: Chemglass
CG-147-30 CG-147-32 CG-147-04 CG-147-01 CG-147-77 CG-147-22 CG-147-21 CG-147-83 CG-147-41 CG-147-76 CG-147-44 CG-147-33 CG-147-24 CG-147-20 CG-147-03 CG-147-23 CG-147-84 CG-147-42 CG-147-75 CG-147-31 CG-147-05 CG-147-34 CG-147-82 CG-147-79 CG-147-43 CG-147-02
80061-180EA 74.16 USD InStock
80061-180 16002-780 80061-168 80061-148 16002-782 89028-494 89028-492 80061-160 80061-182 80061-162 89028-496 80061-184 80061-186 80061-164 80061-166 80061-156 80061-158 80061-170 16002-778 80061-172 80061-150 80061-174 80061-152 80061-154 80061-176 89028-488
Accessories for Reaction Vessels, Chemglass
Reaction Vessels
Flange is formed with a groove in the flat portion of the mating surface to accept an O-Ring.
It also has a ground surface, permitting assembly with either the CG-148 flat flange or CG-149 lid blank using grease.
In the greased application, the O-Ring groove prevents grease from entering the reaction kettle and causing possible contamination.
Outer taper of flange is formed at a 45° angle to accept the CG-141 clamp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty