Would you like to log in again?
Reaction Vessels, Cylindrical, Unjacketed, Large Scale, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1924-34 CG-1924-31 CG-1924-33 CG-1924-35 CG-1924-32 CG-1924-30
16002-832EA 4299.51 USD InStock
16002-832 16002-830 16002-824 16002-822 16002-828 16002-826
Reaction Vessels, Cylindrical, Unjacketed, Large Scale, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Unjacketed large scale process reaction vessels.
  • These flush bottom (zero dead space) valves are designed to prevent solids from collecting in the bottom of the vessel
  • Vessels equipped with the 0-20 mm sealed on valve are supplied complete with a TFE plug having Perfluoro chemically resistant O-rings
  • Vessels are serial numbered and have a graduation scale on both sides showing the approximate volume
The 10, 15, and 20 l vessels have a 200 mm ID flange with an O-ring groove formed in the surface of the flange. The 30 l and 50 l have a 300 mm ID flat ground flange. The 10, 15, and 20 l are available with a choice of a 2" beaded pipe drain that is detachable, or the 0 - 20 mm valve with 3/4" sidearm that is permanently sealed.
Ordering information: Valve, beaded pipe couplings, and adapters must be purchased separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty