Would you like to log in again?
Jacketed Cylindrical Reaction System, Chemglass
Supplier: Chemglass
CG-1965-610 CG-1964-610 CG-1964-610M
16002-918EA 35807.61 USD InStock
16002-918 10170-468 16002-910
Jacketed Cylindrical Reaction System, Chemglass
Laboratory Reactor Systems
Complete reactor system on unique open air, auto-center stand. Electric or air motor.
  • Perfluoro O-rings on 60 mm and 200 mm flanges
  • Heavy-duty lockable casters
  • 5-neck lid with large powder addition port
  • Digital temperature monitor
  • Reactor jacket graduation scale on both sides
  • Removable stainless steel pipe used for supporting side shelf and manifold
  • Support bushings for optional safety shield
Reactor's support frame features oversized rugged stainless steel uprights and heavy-duty PTFE coated aluminum fittings
All models have a 15.3" clearance below valve.
An improved support clamp auto-centers reactor and lid, allowing quick installation and removal of reactor. Stainless steel inlet/outlet adapters available in M16 x 1 and M30 x 1.5 allow connection to cooling/heating circulator. Lower support plate and PTFE ring have a “V” shaped opening, allowing an unobstructed view of bottom valve. Zero dead space detachable drain valve has a knob with laser-etched directional arrows for open and close. The electric motor has a separate controller with digital display.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty