Would you like to log in again?
Glass Baffles for Process Reactors, Chemglass
Supplier: Chemglass
CG-1971-136 CG-1971-122 CG-1971-66 CG-1971-132 CG-1971-74 CG-1971-70 CG-1971-120 CG-1971-72
16003-120EA 395.28 USD InStock
16003-120 16003-098 16003-100 33501-378 16003-104 16003-116 16003-106 16003-118
Glass Baffles for Process Reactors, Chemglass
Stirrers Stirring Tools
Unique glass baffle system for process reactors from 2 to 50 L designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors
  • Available in square or rectangular shapes
  • Open or closed bottom
  • Baffle can be vertically adjusted inside reactor
PTFE joint is offset which permitting baffle to be adjusted up to 1" towards the center of the reactor. The effective baffle length for the reactor size 10 L to 20 L and for 30 L to 50 L are respectively 34 cm and 36 cm.
All baffles are designed with 3/4" precision ground top and lower tubing for use with compression fittings and top jointed adapter.
Delivery information: Complete baffles consist of glass baffle, glass fillted PTFE adapter and lower PTFE compression fitting.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty