Would you like to log in again?
BLEACH-RITE® Disinfecting Spray with Bleach, Current Technologies
Supplier: Current Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BLEACH-RITE®
BRSPRAY32 BRSPRAY128 BRSPRAY16
66025-688CS 310.74 USD InStock
66025-688 16003-740 16003-742
BLEACH-RITE® Disinfecting Spray with Bleach, Current Technologies
Cleaning Agents Disinfectants
Bleach-Rite® Spray is a hospital-grade bleach disinfectant/cleaner approved by the EPA for killing Clostridium difficile Spores (C
Bleach-Rite® Spray is pre-mixed and ready to use at the at the 1:10 dilution (.525% or 5250 ppm) recommended by CDC, OSHA Bloodborne Pathogen Standards and NCCLS. The bleach solution is stabilized for an 18-month shelf life with the expiration date on each bottle and on the outside of each case for inventory management.
Disinfection sites in research, industrial, clinical labs, pharmaceutical and manufacturing plants include centrifuges, chemistry and blood gas analyzers, diagnostic equipment, counters, benchtops, sinks and shelving. Safe to use on plastic, glass, glazed ceramic tile, linoleum, laminated plastic counters, baked enamel, Plexiglas®, and glazed porcelain. For stainless steel, follow equipment manufacturers' directions which may specify that surface be rinsed after disinfection.
Note: Spray will corrode aluminum.
diff spores) on hard, non-porous surfaces in a four-minute contact time. Also approved by the EPA to kill these microorganisms: Thirty-second contact time for Enterococcus faecium (VRE), Streptococcus pyogenes; One-minute contact time for Acinetobacter baumannii, Aspergillus brasiliensis, Bordetella pertussis, Carbapenem-resistant Klebsiella pneumoniae (CRKP), Community acquired methicillin resistant staphylococcus aureus (CA-MRSA), Coronavirus (SARS), Enterobacter aerogenes, Escherichia coli (ESBL), Hepatitis A (HAV), Hepatitis B (HBV), Hepatitis C (HCV), Herpes Simplex (HSV-2), HIV-1 (AIDS), Influenza A H1N1 (swine flu), Influenza A, Norovirus, Poliovirus, Pseudomonas aeruginosa, Respiratory Syncytial (RSV), Rhinovirus, Rotavirus, Salmonella enterica, Methicillin-Resistant Staphylococcus aureus (MRSA), Staphylococcus aureus, Trichophyton mentagrophytes (Athlete's Foot Fungus), Vancomycin-resistant Staphylococcus aureus (VRSA); Two-minute contact time for Canine Parvovirus, Feline Panleukopenia, Tuberculosis (TB), Mycobacterium bovis (BCG); Four-minute contact time for Clostridium difficile Spores (C. diff spores).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty