Would you like to log in again?
Rosetta-gami™ 2 Competent Cells, Expression Strains, MilliporeSigma
Supplier: MilliporeSigma
71350-3 71352-4 71350-3 71351-3 71352-3 71351-3 71351-4 71350-4 71352-4 71351-4 71350-4
16003-960EA 237.88 USD InStock
16003-960 16003-962 16003-952 CA16003-962 CA16003-952 16003-954 16003-956 CA16003-954 CA16003-956 16003-958 CA16003-958
Rosetta-gami™ 2 Competent Cells, Expression Strains, MilliporeSigma
Competent Cells
Rosetta-gami™ 2 host strains combine the advantages of Rosetta 2 and Origami 2 strains to alleviate codon bias and enhance disulfide bond formation in the cytoplasm when heterologoous proteins are expressed in E. coli.
  • Guaranteed efficiency: 2×10⁶ cfu/µg
These trxB/gor mutants are compatible with kanamycin-resistant vectors, and carry the chloramphenicol-resistant pRARE2 plasmid, which supplies seven rare tRNAs.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty