Would you like to log in again?
VWR VistaVision™ Cover Glasses, No. 1
Supplier: VWR International
VWR® VistaVision™
111818 112260 112450VWR 112250 200251 112222 112230 112440 112460 112430 200221 112460A 112240 200181 112525
16004-318CS 296.38 USD InStock
16004-318 16004-308 16004-338 16004-306 16004-328 16004-314 16004-346 16004-310 16004-332 16004-300 16004-330 16004-098 16004-096 16004-094 89218-832
VWR VistaVision™ Cover Glasses, No. 1
Cover Glasses
Microscope cover glasses are made of pure white borosilicate glass with excellent chemical resistance and flatness, and are free of bubbles and scratches.
  • Pure white borosilicate glass fabrication
  • Excellent chemical resistance and flatness
Under ultraviolet light, they emit no disturbing self-fluorescence. Suitable for use in coverslippers. Manufactured in accordance with DIN ISO 8255-1.
Refractive index: 1.525 ±0.0015 ne
Ordering information: Circular and square cover glasses are supplied 100 per two-part box; rectangular cover glasses are supplied 100 per hinged boxes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty